Learn More
GPE Scientific™ J Young Straight Through Taps, Fast Thread Type
GPE Scientific™ J Young Straight Through Taps, Fast Thread Type are made in various stopcock sizes and with different diameter side arms.
Brand:  GPE Scientific™ PTT/10
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15804864
  • 67.20€ / Each
Estimated Shipment: 19-07-2024
Description
  • Fast thread type
  • Available in various stopcock sizes
  • Available with different side arm diameters
Specifications
Specifications
J Young straight through tap with fast thread type
10 mm
ID: 10 mm, OD: 13 mm