Learn More
GPE Scientific™ J Young Spring Clips for Spherical Joints
GPE Scientific™ J Young Spring Clips for Spherical Joints are spring-based clips designed to securely fasten J Young spherical joints.
Marca: GPE Scientific™ JC29
Dettagli aggiuntivi : Weight : 0.05000kg
Descrizione
- Spring clip design
- Securely fasten J Young spherical joints
Specifica
Spring Clips for J Young Spherical Joints | |
28/12 and 28/15 spherical joints |