Learn More
GPE Scientific™ J Young Right Angle Taps, Fine Thread Type
GPE Scientific™ J Young Right Angle Taps, Fine Thread Type are available in various stopcock sizes.
Brand:  GPE Scientific™ POR/20/RA
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15814834
  • 229.00€ / Each
Estimated Shipment: 23-07-2024
Description
  • Fine thread type
  • Available in various stopcock sizes
  • Available with different side arm diameters
Specifications
Specifications
Right angle tap, fine thread
20 mm
ID: 19 mm, OD: 24 mm