Learn More
GPE Scientific™ J Young O-Ring Tap Needle Valve
GPE Scientific™ J Young O-Ring Tap Needle Valve incorporates a PTFE piston sealed as the O-ring with a PTFE needle shut-off point.
Brand: GPE Scientific™ POR/3/NV
Additional Details : Weight : 0.20000kg
Description
- The needle valve incorporates a PTFE piston sealed as the O-ring Tap with a PTFE needle shut-off point.
- Calibrations on the barrel give an indication of flow to enable you to repeat processes.
- Includes a single side arm
- Available with different side arm sizes
Specifications
Needle valve tap with side arm | |
PTFE | |
PTFE piston |
3 mm | |
ID: 3 mm capillary, OD: 6 mm |