Learn More
GPE Scientific™ J Young In Line Taps, Fine Thread
GPE Scientific™ J Young In Line Taps, Fine Thread are made in various stopcock sizes and with various side arm diameters.
Brand:  GPE Scientific™ SPOR/15
Additional Details : Weight : 0.15000kg
 View more versions of this product
Product Code. 15884884
  • 110.00€ / Each
Estimated Shipment: 19-07-2024
Description
  • In-line format tap
  • Available in various stopcock sizes
  • Fine thread type
  • Available with different diameter side arms
Specifications
Specifications
J Young in line tap, fine thread type
15 mm
ID: 15.4 mm, OD: 19 mm