Learn More
GPE Scientific™ J Young Double Oblique Taps, Fine Thread
GPE Scientific™ J Young Double Oblique Taps, Fine Thread are made in various stopcock sizes and with different side arm diameters.
Brand:  GPE Scientific™ POR/D/25
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 15854844
  • 322.00€ / Each
Estimated Shipment: 17-07-2024
Description
  • Fine thread type
  • Available in various stopcock sizes
  • Available with different side arm diameters
Specifications
Specifications
J Young double oblique taps with fine threads
25 mm
ID: 24 mm, OD: 28 mm
Dual taps