Learn More
GPE Scientific™ J Young Double Oblique Taps, Fine Thread
GPE Scientific™ J Young Double Oblique Taps, Fine Thread are made in various stopcock sizes and with different side arm diameters.
Brand:  GPE Scientific™ POR/D/10
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15824844
  • 147.00€ / Each
Estimated Shipment: 17-07-2024
Description
  • Fine thread type
  • Available in various stopcock sizes
  • Available with different side arm diameters
Specifications
Specifications
J Young double oblique taps with fine threads
10 mm
ID: 10 mm, OD: 13 mm
Dual taps