Learn More
Bochem™ Iron Wire Brushes
Product Code. 10631732
Click to view available options
Length (Metric):
270 mm
340 mm
380 mm
420 mm
470 mm
Length (Metric) Brush:
100 mm
120 mm
130 mm
140 mm
170 mm
For Use With (Application):
Flasks
Test tube
Unit Size:
Each
Product Code. 10631732
Brand: Bochem™ 12319
Only null left
Add to basket
Edge
Add to basket
Made with natural bristles. Bochem™ Iron Wire Brushes are used for test tubes and flasks.
  • Iron wire, zincked
  • Natural bristles
  • For test tubes and flasks
Specifications
Product Type Flask Brush
For Use With (Application) Flasks
Length (Metric) 420 mm
Length (Metric) Brush 140 mm
Quantity 1 Ea.