Learn More
Iron(III) chloride hexahydrate, 99+%, extra pure, Thermo Scientific Chemicals
Catalog Number p-4509498
136.80 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
10025-77-1
Cl3FeH12O6
270.29
MFCD00149712
NQXWGWZJXJUMQB-UHFFFAOYSA-K
Ferric chloride hexahydrate
iron(3+) hexahydrate trichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
10025-77-1
Cl3FeH12O6
270.29
MFCD00149712
NQXWGWZJXJUMQB-UHFFFAOYSA-K
Ferric chloride hexahydrate
iron(3+) hexahydrate trichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
TRUSTED_SUSTAINABILITY
Boiling Point 280.0°C to 285.0°C
Melting Point 37.0°C
Color Brown to Yellow
Physical Form Lump
Quantity 1 kg
Assay Percent Range 99% min. (Iodometry)
Linear Formula FeCl3·6H2O
Fieser 01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158
Merck Index 15,4046
Solubility Information Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol,acetone and ether
Formula Weight 270.29
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Iron(III) chloride hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P313-Get medical advice/attention.