missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(Ii) Sulfate Heptahydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
5 g
250 g
1 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 250 g |
Linear Formula | FeSO4 · 7H2O |
IUPAC Name | λ²-iron(2+) heptahydrate sulfate |
Formula Weight | 278.01g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Vapor Pressure | 14.6mmHg (25°C) |
Density | 1.898g/mL at 25°C |
Chemical Name or Material | Iron(Ii) Sulfate Heptahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'