Learn More
Invitrogen™ IP-10 (CXCL10) Human ELISA Development Kit (TMB), PeproTech®, PeproTech®
ELISA
Brand: Invitrogen™ 900-T39
Additional Details : Weight : 0.03500kg
Description
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.Specifications
P02778 | |
3627 | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. |
ELISA | |
CXCL10 | |
RUO | |
Human |