Learn More
Fisherbrand™ Ion Selective Electrode
Combined ion-sensitive electrode with solid membrane interchangeable flat surface where ideally calibration should be carried out as often as possible (Less than once a day).
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
Product Code. 11729718
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • Combined ion-sensitive electrode with solid membrane interchangeable flat surface
  • The ion to be detected passes through the membrane generating a difference of Potential proportional to ionic concentration
  • Analysis time typically less than one minute
  • Sample preparation steps such as filtering or distillation rarely necessary
  • Calibration should be carried out as often as possible (Less than once a day) 
Specifications
Specifications
Ion Selective Electrode