Learn More
SI Analytics™ IoLine pH Combination Electrode
Possess the advantage of a much lower temperature sensitivity and a metal ion free reference system. SI Analytics™ IoLine pH Combination Electrode is for the most demanding measurement tasks.
Brand:  SI Analytics™ 285113900
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11924729
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • Each ID sensor has a unique identification
  • Highest reliability
  • Data exchange between sensor and instrument is fully automatic
  • Several ID sensors can be used with one instrument and one ID sensor can be used with multiple instruments via recognition, without having to re-calibrate
  • Accurate and reliable measurements with sensor specific data GLP at its best
  • Automatic and complete documentation of calibrations and measurements including the electrode used (model and serial number) with date, time and measurement values
Specifications
Specifications
DIN and Banana Plug
Glass
pH Combination Electrode
Sphere
120 mm
Platinum Diaphragm
0 to 14
Iodine/Iodide
Il-PHt-A120-Din-N
12 mm