Learn More
Fisherbrand™ Insert for Midi Boxes
Product Code. 12387167 Shop All Fisherbrand Products
Change view
Click to view available options
Tube Size:
16 x 1.5/16 x 0.5mL
32 x 1.5mL
32 x 2mL
40 x 0.5mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tube Size unitSize
12387167 16 x 1.5/16 x 0.5mL Each
12397157 32 x 1.5mL Each
12317167 32 x 2mL Each
12377157 40 x 0.5mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12387167
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
Manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes; these serve as an ideal solution for short-term storage of radioactive waste or radioisotopes.
  • Microcentrifuge tube beta for FB58447/FB58448
  • 16 x 1.5mL and 16 x 0.5mL tubes 160 x 80 x 40mm
  • Various inserts to fit inside Midi boxes
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
For Use With (Equipment) For Maxi Boxes
Height (Metric) 40mm
Product Type Insert
Tube Size 16 x 1.5/16 x 0.5mL
Width (Metric) 160mm