Learn More
New Brunswick Scientific™ Innova Shaker
Brand:  New Brunswick Scientific™ M1320-0016
Additional Details : Weight : 0.01000kg
Product Code. 11320025
  • 21800.00€ / Each
Estimated Shipment: 26-08-2024
Description
43R console incubated 51mm orbit 25rpm to 300rpm 230V 50Hz 1156mm x 813mm x 1031mm -17°C to +80°C New Brunswick