Learn More
Eppendorf™ Innova™ Benchtop Open Air Shaker
Brand:  Eppendorf™ M1190-0003
Additional Details : Weight : 0.50000kg
Product Code. 10400015
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Specifications
19 mm
± 1 RPM
240 V, 50/60 Hz
50/60 Hz
37 x 35.5 x 14.6 cm
35.5 cm
15 kg
25 to 500 rpm
35 VA
28 cm
14.6 cm
2000