man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Shakers
Rockers and Rotators
11390015
Print
Eppendorf™ Innova™ 43 Incubator Shaker
Brand:
Eppendorf™ M1320-0002
Additional Details :
Weight :
0.01000kg
Product Code.
11390015
19205.00€
/ Each
Quantity
Estimated Shipment: 26-08-2024
Log in
Certificates
Accessories and Spare Parts
Specifications
Orbit
25 mm
Electrical Requirements
230 V, 50 Hz
Holds
6 L
Frequency
50 Hz
Height (Metric)
165.1 cm
Model
Innova 43
Speed Range
25 to 500 rpm
Voltage
230 V
Product Type
Incubator Shaker
Depth (Metric)
81.3 cm
Width (Metric)
115.6 cm
Timer Range
0.01 to 99:59 hr.