Learn More
Fisherbrand™ Class E2 Individual Stainless Steel Wire Calibration Weight with Certificate
Product Code. 15545099 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Weight (Metric):
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
1 unit
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
15545099 10 mg 1 unit
15535099 5 mg 1 unit
15575099 100 mg 1 unit
15565099 50 mg 1 unit
15585099 200 mg 1 unit
15595099 500 mg 1 unit
15555099 20 mg 1 unit
15525099 2 mg 1 unit
15515099 1 mg 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15545099
  • ONLINE EXCLUSIVE PRICE
    71.30€
    60.15€ / Pack of 1
    Save 11.15€
    15% Off
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Wire Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
  • Class E2 calibration weight
  • In compliance with OIML R111
  • Delivered with a calibration certificate guaranteeing traceability
Specifications
Classification E2
Material Stainless Steel
Weight (Metric) 10 mg
Mass 10 mg