Learn More
Fisherbrand™ Individual Stainless Steel Leaf Calibration Weights, Class E2
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Brand: Fisherbrand™
Additional Details : Weight : 0.10000kg
Beschreibung
- Class E2 calibration weight
- In compliance with OIML R111
Spezifikation
E2 | |
Stainless Steel | |
5 mg | |
5 mg |