Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1
Product Code. 15525159 Shop All Fisherbrand Products
Change view
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
1 unit
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) unitSize
15525159 20 g 1 unit
15565139 1 g 1 unit
15565159 50 g 1 unit
15565179 2 kg 1 unit
15585149 10 g 1 unit
15585169 500 g 1 unit
15585189 10 kg 1 unit
15545149 5 g 1 unit
15545169 200 g 1 unit
15525179 1 kg 1 unit
15505189 5 kg 1 unit
15505169 100 g 1 unit
15505149 2 g 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15525159
Only null left
Add to basket
Add to basket
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
  • Class M1 stainless steel calibration weight
  • In compliance with OIML R111
Specifications
Classification M1
Material Stainless Steel
Weight (Metric) 20 g
Mass 20 g
Shape Cylindrical