Learn More
Honeywell Safety Products™ Indirect Vent, LG20 Safety Goggles
Ensure high-level protection against flying particles, liquid droplets, mists and sprays. Honeywell Safety Products™ Indirect Vent, LG20 Safety Goggles are lightweight, effective and adjustable.
Brand: Honeywell Safety Products™ 1005509
Additional Details : Weight : 0.09000kg
Description
- Impact resistant polycarbonate lens for excellent protection against flying particles, coarse dusts, liquid droplets, mists and sprays.
- Lightweight design.
- Soft seal and easily adjustable headband fits a wide variety of workers.
- Fog-Ban lens.
- Economy goggles with good impact resistance (120 m/s) and large field of vision.
- Indirect ventilation (4 vents)
Specifications
Impact-resistant | |
Safety glasses | |
1 Ea. | |
Polycarbonate |
EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC0835, EC Attestation Number EC0835 | |
Automotive, Building, Construction, Food, Industrial Cleaning, Laboratory and Maintenance industries. | |
75 to 79 g |