Learn More
Invitrogen™ IL-17 E Human ELISA Development Kit (TMB), PeproTech®, PeproTech®
ELISA
Brand:  Invitrogen™ 900-T234
Additional Details : Weight : 0.03500kg
Product Code. 17888915
  • 330.00€ / Pack of 10
Estimated Shipment: 18-06-2024
Description
Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
Specifications
Specifications
Q9H293
64806
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
ELISA
IL25