Learn More
IKA™ R 2723 Telescopic Stand
Shop All IKA Products
Click to view available options
Depth (Metric):
420 mm
Height (Metric):
1010 mm
Unit Size:
Each
Description
Provides optimum stability required for larger, heavier instruments and attachments, for example with rheological measurements using overhead stirrers. The stand has an adjustment screw which can be used to compensate for an uneven laboratory table surface.
Specifications
Specifications
| Depth (Metric) | 420 mm |
| Height (Metric) | 1010 mm |
| Quantity | 1 Ea. |
| Type | Telescopic Stand |
| Width (English) | 18 in. |
| Width (Metric) | 460 mm |