missing translation for 'onlineSavingsMsg'
Learn More

Proteintech IHCeasy VWF Ready-To-Use IHC Kit

Ready-To-Use IHC Kit

Brand:  Proteintech KHC0209-50T

Additional Details : Weight : 0.70000kg

Product Code. 17600277

  • 389.00€ / 1 set
Estimated Shipment: 27-05-2024
to see stock.
Add to basket

Description

Description

KHC0209 is a ready-to-use IHC kit for staining of VWF (von Willebrand factor). The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications

Specifications

Monoclonal
50 Tests
Mouse
  • Antigen Retrieval Buffer(100 mL, 50 x)
  • Washing Buffer(100 mL x2, 20 x)
  • Blocking Buffer(5 mL, RTU)
  • Rabbit Polyclonal Primary Antibody(5 mL, RTU)
  • Polymer-HRP-Goat anti-Mouse Secondary Antibody(5 mL, RTU)
  • Chromogen Component A(0.2 mL, RTU)
  • Chromogen Component B(4 mL, RTU)
  • Signal Enhancer(5 mL, RTU)
  • Counter Staining Reagent(5 mL, RTU)
  • Mounting Media(5 mL, RTU)
  • Datasheet (1 Copy)
  • Manual(1 Copy)
Human
Immunohistochemistry (IHC)
All the reagents are stored at 2°C to 8°C. The kit is stable for 6 months from the date of receipt.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers