Learn More
Masterflex™ Hose Clamp for Peristaltic Pump
Reusable white actal copolymet clamps will not rust of corrode and stay in place once positioned
52.55€ - 74.60€
Specifications
Material | Acetal Copolymer |
---|---|
For Use With (Equipment) | Hose or tubing |
Color | White |
Product Code | Brand | Diameter (English) Inside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inside | Price | Quantity & Availability | |||||
12499650
|
Masterflex™
06832-03 |
Min. I.D. 0.307 in., max. I.D. 0.342 in. |
53.35€
Pack of 100 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
12419660
|
Masterflex™
06832-06 |
Min. I.D. 0.449 in., max. I.D. 0.511 in. |
53.20€
Pack of 100 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12439660
|
Masterflex™
06832-10 |
Min. I.D. 0.569 in., max. I.D. 0.65 in. |
57.35€
Pack of 100 |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12449660
|
Masterflex™
06832-14 |
Min. I.D. 0.702 in., max. I.D. 0.801 in. |
61.95€
Pack of 100 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
12469660
|
Masterflex™
06832-20 |
Min. I.D. 0.859 in., max. I.D. 0.989 in. |
61.95€
Pack of 100 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
12479660
|
Masterflex™
06832-22 |
Min. I.D. 0.928 in., max. I.D. 1.072 in. |
74.60€
Pack of 100 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
12489660
|
Masterflex™
06832-32 |
Min. I.D. 1.218 in., max. I.D. 1.405 in. |
52.55€
Pack of 50 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
Description
- Acetal copolymer
- Suitable for continuous use from ambient temperature to 93°C
- Non-conducting and stable in a wide variety of chemicals
- Clamps stay in place once positioned
- Position clamp and press jaws together with pliers
Specifications
Acetal Copolymer | |
Hose or tubing | |
White |