Learn More
Sperian™ Honeywell™ ARMAMAX AX™ Clear Lens Safety Goggles
Product Code. 12970405
Change view
Click to view available options
Lens Coating:
Hardcoat
Uncoated
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Coating Unit Size
12970405 Hardcoat Each
11847221 Hardcoat Pack of 10
11857221 Uncoated Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12970405 Supplier Sperian™ Supplier No. 1002221
  • ONLINE EXCLUSIVE PRICE
    12.30€
    8.68€ / Pack of 1
    Save 3.62€
    29% Off
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
Offering high levels of comfort and support. Sperian™ Honeywell™ ARMAMAX AX™ Clear Lens Safety Goggles fit comfortably over prescription eyewear.
  • Practical and durable
  • Frosted side panels and browguard sections reduce glare/reflections
  • Curved temples and a raised nose bridge offer a precise fit
  • Available with or without hard lens coating
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001 / 2000, EN 166 1F
Lens Coating Hardcoat
Lens Tint Clear
Product Type Eye and Face Protection Spectacles
For Use With (Application) Over-the-glass spectacle, General use and protection against impact (45m/s), Mechanic work: Grinding, turning, milling
Frame Color Transparent
No. per Pack 10/Pk.
Lens Material Polycarbonate