Learn More
Sperian™ Honeywell™ A700 Series Clear Hard Coat Safety Glasses
Providing rock solid protection at a rock bottom price. Sperian™ Honeywell™ A700 Series Clear Hard Coat Safety Glasses provide a stylish fit for the everyday wearer.
Brand: Sperian™ 1015361
Additional Details : Weight : 0.02700kg
Description
- Affordably priced and economical for your business
- Lightweight design with sport temples and soft temple tip pads
- Moulded universal nose bridge
- Provides a secure, wraparound fit for all-day comfort
- 8-base polycarbonate abrasion-resistant lenses
- Provides a 180° field of distortion-free vision
Specifications
Scratch-resistant | |
Eye and Face Protection Spectacles | |
Clear | |
Polycarbonate |
Clear | |
General use and protection against impact (45m/s), Well suited for Indoor applications such as light manufacturing. | |
Polycarbonate |