Learn More
Cytiva HiTrap™/HiPrep™ Column Accessories
81.00€
Product Code | Brand | Product Type | Quantity | For Use With (Application) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | For Use With (Application) | Price | Quantity & Availability | |||
11802200
|
Cytiva
11-0004-64 |
Sto p plug | 5 Pack | HiTrap column (sealing botto m of column) |
81.00€
Pack of 5 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||
11726964
|
Cytiva
28-4010-81 |
Connecto r | 8 Pack | HiTrap and HiPrep column (connecting column to AKTA™ design systems) |
81.00€
Pack of 8 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||
Specifications
Polypropylene | |
Column Accesory |