Learn More
Honeywell™ HIKE (i)XTREM S3 Shoes
Honeywell™ HIKE (i)XTREM S3 Shoes are constructed of water-resistant full grain leather.
Brand: Honeywell™ 6246201-47/7
Additional Details : Weight : 2.25000kg
Beschreibung
- Water resistant grain full leather
- Ergonomic collar and padded tongue for an optimized comfort
- Gusset to avoid water and dust penetration
- Breathable 3D lining
- Overcap
- Toe cap metal free
- Metal free antiperforation insert
- Injected PU / Nitrile sole
- (i)NTELLIGENT wear control
- EN ISO 20345:2011 S3 HRO HI CI SRC
Spezifikation
EN ISO 20345:2011 S3 HRO HI CI SRC | |
Leather | |
Polyurethane and Nitrile |
Brown, Black | |
47 | |
Metal-free |