Learn More
UVP High-Performance Transilluminator (25W) 2Uv Single Intensity
Brand:  UVP 95-0432-02
Additional Details : Weight : 10.24000kg
Product Code. 12853938
  • 2225.00€ / Each
Estimated Shipment: 26-07-2024
Description
Electronic ballast stainless steel filter assembly 302/365nm wavelength 230V 50Hz purple base 200mm x 400mm filter size UVP