Learn More
Abnova™ HER4/CEN2p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0024.100uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16033015
  • 1475.00€ / 100µL
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
Includes: DAPI Counterstain (1500ng/mL) 125μL
Description
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
Store at 4°C in the dark.
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
2q33-q34, 2p11.2
HER4/CEN2p FISH Probe
FITC, Texas Red
DAPI Counterstain (1500 ng/mL ) 250 uL
HER4, CEN2p