Learn More
Greiner Bio-One Heavy Designed Polystyrene Vented Petri Dishes
Used for easy stacking. Greiner Bio-One Heavy Designed Polystyrene Vented Petri Dishes is designed for better heat resistance using hot agar.
189.00€
Product Code | Brand | Diameter (Metric) | Height (Metric) | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Height (Metric) | Sterility | Price | Quantity & Availability | |||
10567381
|
Greiner Bio-One
633102 |
94 mm | 16 mm | Non-sterile |
189.00€
Pack of 480 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
Description
- Vented for better gas exchange
- Heavy design for heat resistance using hot agar
- Manufactured from crystal polystyrene for optimal analysis by microscopy
Specifications
Polystyrene | |
Round | |
Yes |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.