Learn More
Hamilton™ Removable Needle to Luer Adapters
These adapters convert a Hamilton specific termination to an industry standard luer connection.
Brand: Hamilton™ 35083
Additional Details : Weight : 0.00100kg
Description
These adapters convert a Hamilton specific termination to an industry standard luer connection.
- Compatible with SampleLock or Removable Needle syringes 250μL and larger
- Internal diameter is 1mm making it possible to spike samples with an internal standard using a 26 gauge needle or smaller
- Male luer lock connection
Specifications
Male Luer-lok Hub Adapter, for 10 to 33 ga. Needles | |
1/Pk. | |
Male Luer Lock | |
Removable Needle Syringe (250μL and Larger) |