Learn More
Gilac™ HACCP Food Contact Container
HACCP Food Contact Container is ideal for storage, food preparation, and transporting foodstuffs.
Brand:  Gilac™ G613228
Additional Details : Weight : 2.47000kg
 View more versions of this product
Product Code. 17978511
  • 35.32€ / Each
Estimated Shipment: 05-06-2024
Description
  • HACCP round container.
  • Food contact certified.
  • For storage, food preparation and transporting foodstuffs.
  • Highly durable.
  • Reinforced bottom with anti-wear skirt.
  • Reinforced handles to prevent distortion during carrying.
  • Optional, choice of two lids.
  • External Diameter includes handles.
  • Width corresponds to diameter without handles.
Specifications
Specifications
HACCP Food Container
Blue
50 L
Polyethylene
13.2 gal.