Learn More
Fisherbrand™ Microscope Slides with Ground Edges
Product Code. 12383118 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Thickness (Metric):
0.8 to 1 mm
1 to 1.2 mm
1.2 to 1.5 mm
Frosted:
Not Frosted
Twin Frosted
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Frosted Unit Size
12383118 1 to 1.2 mm Not Frosted Pack of 50
11562203 0.8 to 1 mm Twin Frosted Pack of 50
12373118 0.8 to 1 mm Not Frosted Pack of 50
11572203 1 to 1.2 mm Twin Frosted Pack of 50
12393118 1.2 to 1.5 mm Not Frosted Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12383118
  • ONLINE EXCLUSIVE PRICE
    4.11€
    3.00€ / Pack of 50
    Save 1.11€
    27% Off
In Stock
Add to basket
Edge
Add to basket
Wrapped in cellophane for improved cleanliness. These Microscope Slides with Ground Edges are washed and polished glass slides with 90° ground edge.
  • Glass 76 mm x 26 mm
  • Available in fully clear glass or frosted at one end
  • Ground on all four edges
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
Edges 90°, Ground
Frosted Not Frosted
Material Glass
Precleaned Yes
Thickness (Metric) 1 to 1.2 mm
Type Plain Microscope Slide
Color Clear
Quantity 50 Pk.