Learn More
Grant Instruments™ Grant Bio™ Adjustable Bar for Orbital Platform Shaker
Keeps flasks in place and secure while shaking and rotating. For use with Universal Platform and Compact Shaker-Incubator
Brand: Grant Instruments™ HB-200
Additional Details : Weight : 0.06300kg
Description
- For use with Universal Platform or Compact Shaker-Incubator
Specifications
Adjustable Bar | |
PUP-12 universal platform in conjunction with ES-20 Shaker-Incubator | |
1 Ea. |