Learn More
Fisherbrand™ Grade 7 Cellulose Fiber and Cotton Linter Extraction Thimble
Product Code. 11724043 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Inside:
16 mm
19 mm
22 mm
25 mm
26 mm
27 mm
28 mm
30 mm
33 mm
34 mm
35 mm
37 mm
41 mm
43 mm
58 mm
70 mm
80 mm
Length (Metric):
40 mm
60 mm
80 mm
90 mm
94 mm
100 mm
118 mm
120 mm
123 mm
130 mm
150 mm
170 mm
200 mm
240 mm
250 mm
Unit Size:
Pack of 25
29 product options available for selection
Product selection table with 29 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Length (Metric) Unit Size
11724043 19 mm 90 mm Pack of 25
11764526 16 mm 100 mm Pack of 25
11734536 19 mm 40 mm Pack of 25
11774526 22 mm 60 mm Pack of 25
11754043 25 mm 100 mm Pack of 25
11734043 22 mm 80 mm Pack of 25
11774043 26 mm 60 mm Pack of 25
11744043 25 mm 80 mm Pack of 25
11784526 27 mm 80 mm Pack of 25
11794043 28 mm 100 mm Pack of 25
11542103 28 mm 120 mm Pack of 25
11784043 28 mm 80 mm Pack of 25
11714053 30 mm 100 mm Pack of 25
11724536 30 mm 150 mm Pack of 25
11704053 30 mm 80 mm Pack of 25
11734053 33 mm 100 mm Pack of 25
11744536 33 mm 118 mm Pack of 25
11724053 33 mm 80 mm Pack of 25
11714536 33 mm 94 mm Pack of 25
11794526 34 mm 130 mm Pack of 25
11764536 35 mm 150 mm Pack of 25
11744053 37 mm 130 mm Pack of 25
11754053 41 mm 123 mm Pack of 25
11704536 41 mm 150 mm Pack of 25
11754536 43 mm 123 mm Pack of 25
11774536 58 mm 170 mm Pack of 25
11784536 70 mm 240 mm Pack of 25
11794536 80 mm 200 mm Pack of 25
11704546 80 mm 250 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
29 options
Product Code. 11724043
  • ONLINE EXCLUSIVE PRICE
    46.10€
    41.66€ / Pack of 25
    Save 4.44€
    9% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
Suitable for all extractors systems.
  • 100% High pure cotton linters fibers
  • Neutral and fat free
  • High mechanical strength
  • Excellent retention capacity
Applications:
  • Extraction of sulphur
  • Extraction of the content of free lipids in food
  • Determination of dust and tar in industrial air and gases
  • Lacquer, binder analysis in paint colours
  • Control of emission smokes in industrial chimneys
  • Gravimetric analysis of dust in gases
Specifications
Diameter (Metric) Inside 19 mm
Material High Purity Cellulose Fibers and Cotton Linters
For Use With (Application) Extraction of Sulfur in gaseous masses, determination of dust and tar in industrial air and gases
Grade 7
For Use With (Equipment) Suitable for all extractors systems
Length (Metric) 90 mm
Product Type Extraction Thimble