Learn More
Fisherbrand™ Grade 122 Cellulose General Purpose Filter Paper
Very fast filtration velocity. Fisherbrand™ Grade 122 Cellulose General Purpose Filter Paper is used for all general works in laboratories.
Brand:  Fisherbrand™
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11708752
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Very fast filtration velocity
  • Used for general works, hospitals, research laboratories
Applications:
  • Chemical industries, pharmaceutical, cosmetic, food and beverage
  • Specifications
    Specifications
    Disc
    17 to 30 μm
    For all general works in laboratories
    100/Pk.
    130 mm
    Cellulose
    17 to 30 μm
    122
    0.17 mm
    130 mm