Learn More
Fisherbrand™ Grade 1105 Cellulose Technical Filter Paper
Suitable for filtration of large volume of liquid.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11760026
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Very thick smooth paper
  • Wet strengthened
  • With high separation capacity
Specifications
Specifications
Disc
3 to 6 μm
For dyes, Ether, Alcoholic solutions, Eaux de Cologne, salted solutions, vegetal extracts, hair solutions, perfumes, tinctures, saccharine juices, vinegar
100 Pack
185 mm
Cellulose
3 to 6 μm
1105
0.39 mm
185 mm