missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 10 Smooth Surface Folded Filter Qualitative Filter Papers

Used for routine analyses like clarification and determination of substances

Brand:  Sartorius™ FT-4-352-185

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10460462

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »


Description

Description

  • Made of refined pulp and cotton linters with an >95% α-cellulose content
  • Ash content between 0.1–0.15% (grade 100/N <0.1%)
  • Wet-strengthened
  • Supplied in rolls, sheets, discs and folded filters as well as customer-specific cuts
Specifications

Specifications

Folded
7.5μm/sec.
10
0.33 mm
185 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
100/Pk.
Qualitative Filter Paper
185 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers