Learn More
3M™ GoggleGear™ Protective Goggle 3000 Series, TPV Shroud, Clear Scotchgard™ AF Lens, Premium Cloth Strap
Description
- 3M™ GoggleGear™ Protective Goggle 3000 Series features a full-size design with wrap-around lens for improved field of vision, and slotted nose bridge for a more comfortable fit.
- Choose from models with anti-fog or 3M™ Scotchgard™ Anti-Fog coating.
- The GoggleGear™ 3000 is available with indirect venting for enhanced air circulation and anti-fog performance, and is 3, protection against droplets, 4, protection against large particles, and 6, protection against streams of liquid, rated.
- Includes either an adjustable polyester cloth strap, or easy to clean adjustable neoprene strap, as well as an optimized shroud design with slots to accommodate many prescription eyewear.
- Acetate lens model meets the chemical resistance requirement of ISO 16321, helping protect workers from chemical hazards.
Specifications
Specifications
| Lens Coating | Anti-fog |
| Color | Black |
| Lens Tint | Clear |
| Product Type | Anti-fog Goggles |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | TPV |
| UV Protection | None |
| Frame Type | Full Frame |