Learn More
Rogo Sampaic™ Glass Slides
Brand:  Rogo Sampaic™ BPB032
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11852350
  • 9.81€ / Pack of 50
Estimated Shipment: 18-07-2024
Specifications
Cut
76 mm
Blue
76 x 26 mm
Glass
26 mm
50 Pack