Learn More
Cole-Parmer™ Glass Sample Vials
Glass Sample Vials maintain PH level and eliminate ion leaching.
157.82€ - 292.00€
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15274393
|
Cole-Parmer™
08922-44 |
12 mL |
292.00€
Pack of 200 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15192944
|
Cole-Parmer™
08922-39 |
40 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- Vials are manufactured from 33 expansion borosilicate clear glass (Type 1, Class A).
- Choose from a polypropylene cap with a PTFE-lined foam backing or a polypropylene cap with PTFE-lined silicone septa.
- Vials, Caps, and septa are shrink-wrapped separately.
Specifications
Borosilicate Glass (33 Expansion) | |
8 mm |