Learn More
Sartorius Stedim Biotech™ Glass Microfiber Filters, Grade MGC
Binder-free glass microfiber filters for analytical and gravimetric analyses and prefiltering.
Brand: Sartorius Stedim Biotech™ FT-2-1103-460570
Additional Details : Weight : 1.96000kg
Description
- Binder-free
- Suitable for the following:
- Analysis of suspended solids in wastewater according to EN 872
- Liquid filtration
- Prefiltration
- Clarification
- Analytical sample preparation
- Particle analysis
- Non-sterile, bulk packed
Specifications
54 g/m2 | |
Sheet | |
1.2 μm | |
335 mL/min. | |
Hydrophilic | |
0.26 mm |
White | |
Binder-free Glass Microfiber | |
Non-sterile | |
MGC | |
100/Pk. | |
460 x 570 mm |