Learn More
Merck Glass Funnel for Vacuum Filtration
Description
Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration
Specifications
Specifications
| Diameter (Metric)Top | 47 mm |
| Capacity (Metric) | 300 mL |
| Material | Borosilicate Glass |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Description | Ground glass seal |