Learn More
- Home
- Products
- Flasks
- Volumetric Flasks
- Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Product Code. 11782142
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
34/35 NS Standard Ground
45/40 NS Standard Ground
7/16 NS Standard Ground
7/16 NS Standard Taper Ground
Tolerance:
±0.025 mL
±0.04 mL
±0.06 mL
±0.08 mL
±0.1 mL
±0.15 mL
±0.25 mL
±0.4 mL
±0.6 mL
±1.2 mL
±2 mL
0.025 mL
Unit Size:
Each
Pack of 2
20 product options available for selection
| Product Code. | Capacity (Metric) | Standard Taper Joint | Tolerance | Unit Size |
|---|---|---|---|---|
| 11782142 | 200 mL | 14/23 NS Standard Ground | ±0.15 mL | Pack of 2 |
| 10218211 | 10 L | 45/40 NS Standard Ground | ±2 mL | Each |
| 10689451 | 1000 mL | 24/29 NS Standard Ground | ±0.6 mL | Each |
| 10522891 | 1000 mL | 24/29 NS Standard Ground | ±0.4 mL | Each |
| 11780964 | 100 mL | 12/21 NS Standard Ground | ±0.1 mL | Pack of 2 |
| 11760964 | 10 mL | 10/19 NS Standard Ground | ±0.04 mL | Pack of 2 |
| 11708071 | 10 mL | 7/16 NS Standard Ground | ±0.025 mL | Pack of 2 |
| 10561161 | 2000 mL | 29/32 NS Standard Ground | ±0.6 mL | Each |
| 11718071 | 20 mL | 10/19 NS Standard Ground | ±0.04 mL | Pack of 2 |
| 11792142 | 250 mL | 14/23 NS Standard Ground | ±0.15 mL | Pack of 2 |
| 11816993 | 25 mL | 12/21 NS Standard Ground | ±0.06 mL | Pack of 2 |
| 11728071 | 25 mL | 10/19 NS Standard Ground | ±0.04 mL | Pack of 2 |
| 10244712 | 20 mL | 12/21 NS Standard Ground | ±0.04 mL | Pack of 2 |
| 11702152 | 500 mL | 19/26 NS Standard Ground | ±0.25 mL | Pack of 2 |
| 10646111 | 5 L | 34/35 NS Standard Ground | ±1.2 mL | Each |
| 11770964 | 50 mL | 14/23 NS Standard Ground | ±0.08 mL | Pack of 2 |
| 11750964 | 5 mL | 10/19 NS Standard Ground | ±0.04 mL | Pack of 2 |
| 11738071 | 50 mL | 12/21 NS Standard Ground | ±0.06 mL | Pack of 2 |
| 11772142 | 100 mL | 14/23 NS Standard Ground | ±0.1 mL | Pack of 2 |
| 11782412 | 5 mL | 7/16 NS Standard Taper Ground | 0.025 mL | Pack of 2 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
Description
- With NS ground and polyethylene stopper
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 1042 |
| Closure Material | Polyethylene |
| Capacity (Metric) | 200 mL |
| Material | Borosilicate Glass, Duran Glass |
| Height (Metric) | 210 mm |
| Limit of Error | ±0.15 mL |
| Standard Taper Joint | 14/23 NS Standard Ground |
| Tolerance | ±0.15 mL |
| Graduated | Yes |
| Class | A |