Learn More
Gilson™ TRACKMAN™ Duo-plate Pipetting Tracker Accessory, Microtube Holder
Optional tube holder accessory for the TRACKMAN™ Duo-plate Pipetting Tracker.
Brand:  Gilson™ F70105
Additional Details : Weight : 0.01000kg
Product Code. 12376112
  • 31.45€ / Each
Estimated Shipment: 19-07-2024
Description
  • 0.5 mL, 1.5 mL and 2 mL microtube-holder
  • Vessel capacity: 16 microtubes
Specifications
Specifications
Microtube Holder
0.5 mL, 1.5 mL, 2 mL Tube
Optional tube holder accessory