Learn More
Gilson™ PIPETMAN™ EXPERT™ Universal Fit Gel Loading Pipet Tips: Sterile
Flat or round tip styles are available
Brand: Gilson™ F1733382
Additional Details : Weight : 1.78000kg
Description
- Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
- Features graduations at varying volumes allowing for quick-checks when aspirating samples
Specifications
1 to 200 μL | |
Clear | |
8.255cm | |
Yes | |
Polypropylene |
Sterile | |
12.75 in. | |
Flat | |
Pipetman EXPERT | |
5μL |