Learn More
Gilson™ Accessory for PIPETMAN™ Complete Pipetting System- Coloris™ Colored Identifi cation Clips
GLP-friendly, brightly colored clips made of polyacetal are used for identifying Pipetteman™ and Neo™ pipets
20.70€
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
10530923
|
Gilson™
F161301 |
Assorted |
20.70€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10162902
|
Gilson™
F161303 |
Yellow |
20.70€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10666843
|
Gilson™
F161305 |
Blue |
20.70€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10152952
|
Gilson™
F161304 |
Green |
20.70€
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
10337662
|
Gilson™
F161306 |
White |
20.70€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10626463
|
Gilson™
F161302 |
Red |
20.70€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
- Brightly colored clips
- Made of polyacetal
- Used for identifying Pipetteman and Neo pipettes
Specifications
Polyacetal |