Learn More
GE Healthcare Amersham™ Hybond™-N Membranes
Use for high sensitivity applications in DNA and RNA blotting. GE Healthcare Life Sciences™Amersham™ Hybond™-N Membranes is a strong supported membrane that is inherently hydrophilic and requires no pre-wetting.
266.00€ - 394.00€
| Product Code | Brand | Description | Format | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Description | Format | Price | Quantity & Availability | ||||
| 10217654 |
GE Healthcare
RPN203N |
20cm x 3m | Roll |
282.00€ Each |
|
10074194 |
GE Healthcare
RPN303N |
30cm x 3m | Roll |
394.00€ Each |
|
11768338 |
GE Healthcare
RPN2020N |
20 x 20cm | Sheet |
282.00€ Pack of 10 |
|
11778398 |
GE Healthcare
RPN82N |
8.2cm | Disc |
279.00€ Pack of 50 |
|
10361475 |
GE Healthcare
RPN1520N |
15 x 20cm | Sheet |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
Description
- Binding capacity for nucleic acids up to 600μm/cm2
- Inherently hydrophilic and requires no prewetting
- Nucleic acids can be crosslinked using UV light
Specifications
| Up to 600μg/cm2 | |
| Nylon, Neutral |